Order Entry
Belgium
ContactUsLinkComponent
Iridium standard solution, 100 mg/L Ir in 10% hydrochloric acid standard for ICP-MS
Iridium standard solution, 100 mg/L Ir in 10% hydrochloric acid standard for ICP-MS
Catalog # ACCUICPMS26H001X1
Supplier:  AccuStandard
CAS Number:  
Iridium standard solution, 100 mg/L Ir in 10% hydrochloric acid standard for ICP-MS
Catalog # ACCUICPMS26H001X1
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    10% HCl
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Iridium
  • Symbol:
    Ir

Specifications

About this item