Order Entry
Belgium
ContactUsLinkComponent
Iron standard solution, 10000 mg/L Fe in 10% nitric acid (from Fe(NO₃)₃) Certipur® standard for ICP, Supelco®
Iron standard solution, 10000 mg/L Fe in 10% nitric acid (from Fe(NO₃)₃) Certipur® standard for ICP, Supelco®
  1.70376.0100
 :  Merck
 :  
Iron standard solution, 10000 mg/L Fe in 10% nitric acid (from Fe(NO₃)₃) Certipur® standard for ICP, Supelco®
  1.70376.0100
 :  Merck
 :  

 

  • Concentration:
    10000 mg/L
  • Matrix:
    10% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Iron
  • Symbol:
    Fe

 

 

 : Traceable to SRM from NIST