Calcium standard solution, 1000 mg/L Ca in 5% nitric acid (from CaCO₃), standard for ICP, Specpure®
Molecular Formula: CaCO₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085314
: Traceable to NIST
- MDL No.:MFCD00085314
Frequently Bought Together