Order Entry
Belgium
ContactUsLinkComponent
Pesticide reference standard, Aldrin 100 µg/ml in n-nonane, CIL
Pesticide reference standard, Aldrin 100 µg/ml in n-nonane, CIL
Catalog # PROOCILULM74411.2
CAS Number:  
Pesticide reference standard, Aldrin 100 µg/ml in n-nonane, CIL
Catalog # PROOCILULM74411.2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    309-00-2
  • Description:
    Aldrin 100 µg/ml in n-nonane
  • Formula:
    C₁₂H₈Cl₆
  • Description:
    Aldrin

Specifications

About this item