Order Entry
Belgium
ContactUsLinkComponent
 
Rhodium standard solution, 10 mg/L Rh in 20% hydrochloric acid (from RhCl₃), standard for ICP, Specpure®
  45320.AE
 :  
Rhodium standard solution, 10 mg/L Rh in 20% hydrochloric acid (from RhCl₃), standard for ICP, Specpure®
  45320.AE
 :  

 

  • Concentration:
    10 mg/L
  • Matrix:
    20% HCl
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • MDL No.:
    MFCD00011201
  • Element:
    Rhodium
  • Symbol:
    Rh

 

 

Molecular Formula: RhCl₃ in 2% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011201

 : Traceable to NIST