Order Entry
Belgium
ContactUsLinkComponent
 
Mercury standard solution, 1,000 mg/L Hg in 0.5 M nitric acid standard for AAS
  REGEAAHGH
 :  REAGECON
 :  
Mercury standard solution, 1,000 mg/L Hg in 0.5 M nitric acid standard for AAS
  REGEAAHGH
 :  REAGECON
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Mercury
  • Symbol:
    Hg