Order Entry
Belgium
ContactUsLinkComponent
Mercury standard solution, 100 mg/L Hg in dil. nitric acid standard for ICP-MS
Mercury standard solution, 100 mg/L Hg in dil. nitric acid standard for ICP-MS
Catalog # ACCUICPMS34N001X1
Supplier:  AccuStandard
CAS Number:  
Mercury standard solution, 100 mg/L Hg in dil. nitric acid standard for ICP-MS
Catalog # ACCUICPMS34N001X1
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Mercury
  • Symbol:
    Hg

Specifications

About this item