Order Entry
Belgium
ContactUsLinkComponent
Indium standard solution, 100 mg/L In in dilute nitric acid standard for ICP-MS
Indium standard solution, 100 mg/L In in dilute nitric acid standard for ICP-MS
  ACCUICPMS25N001X1
 :  
Indium standard solution, 100 mg/L In in dilute nitric acid standard for ICP-MS
  ACCUICPMS25N001X1
 :  

 

  • Concentration:
    100 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Indium
  • Symbol:
    In