Gallium standard solution, 1000 mg/L Ga in 5% nitric acid with hydrochloric acid (max. 1%) (from Ga), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular Formula: Ga in 5% HNO₃/tr. HCl
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134045
Certifications: Traceable to NIST
Specifications
- MDL No.:MFCD00134045
Specifications
Related Information
Frequently Bought Together