Order Entry
Belgium
ContactUsLinkComponent
Cobalt standard solution, 10 mg/L Co in 5% nitric acid (from Co), standard for ICP, Specpure®
Cobalt standard solution, 10 mg/L Co in 5% nitric acid (from Co), standard for ICP, Specpure®
Catalog # 45252.AE
CAS Number:  
Cobalt standard solution, 10 mg/L Co in 5% nitric acid (from Co), standard for ICP, Specpure®
Catalog # 45252.AE
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • MDL No.:
    MFCD00010935
  • Element:
    Cobalt
  • Symbol:
    Co

Specifications

About this item

Molecular Formula: Co in 2% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00010935

Certifications: Traceable to NIST