
Hydrochloric acid 1.8 mol/l (1.8 N)
Catalog # VWRS5327.9010
Supplier: VWR Collection
CAS Number:
Specifications
- Pack type:Plastic container
- Pk:10 L
- Seq:5214
Specifications
About this item
:
ADR:8,II
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS05
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324