Order Entry
Belgium
ContactUsLinkComponent
 
Herbicide reference standard, Desisopropyl atrazine 10 µg/ml in acetonitrile
  PROONE7054A
 :  
Herbicide reference standard, Desisopropyl atrazine 10 µg/ml in acetonitrile
  PROONE7054A
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    1007-28-9
  • Description:
    Desisopropyl atrazine 10 µg/ml in acetonitrile
  • Formula:
    C₅H₈ClN₅
  • Description:
    Desisopropyl atrazine