Order Entry
Belgium
ContactUsLinkComponent
2,2,4-Trimethylpentane ≥99.5%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Flammable
Health
Irritant
Environment
2,2,4-Trimethylpentane ≥99.5%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # 6043-25
CAS Number:  
Flammable
Health
Irritant
Environment
2,2,4-Trimethylpentane ≥99.5%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # 6043-25
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • ACS - Assay ((CH₃)₃CCH₂CH(CH₃)₂) (by GC, corrected for water):
    ≥99.5 %
  • ACS - Color (APHA):
    ≤10
  • ACS - Residue after Evaporation:
    ≤0.0003 %
  • ACS - Sulfur Compounds (as S):
    ≤0.005 %
  • ACS - Water (H₂O)(by Karl Fischer titrn):
    ≤0.02 %
  • ACS - Water-Soluble Titrable Acid (meq/g):
    ≤0.0003
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 205 nm:
    ≤1.00
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm:
    ≤1.00
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm:
    ≤0.20
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm:
    ≤0.10
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm:
    ≤0.04
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 250 - 400 nm:
    ≤0.01

Specification Test Results

About this item

 :
 :114.23 g/mol
Merck Index:13,05212
Boiling Pt:99.2 °C (1013 hPa)
Melting Pt:–107 °C
Density:0.692 g/cm³ (20 °C)
Flash Pt:–12 °C
Storage Temperature:Ambient
EINECS:208-759-1
UN:1262
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:540-84-1
Formula:(CH₃)₂CHCH₂C(CH₃)₃
MDL Number:MFCD00008943