Order Entry
Belgium
ContactUsLinkComponent
1-Butanol for HPLC
Irritant
Corrosive
Flammable
1-Butanol for HPLC
   Sustainable Options Available
Certificates
undefined
Irritant
Corrosive
Flammable
1-Butanol for HPLC
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.05 %
  • GC:
    ≥99.7 %
  • UV:
    1 cm cell vs HPLC-grade water
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.0003 %
  • Acidity:
    ≤0.0002 meq/g

Specification Test Results

Related Information

Frequently Bought Together