Order Entry
Belgium
ContactUsLinkComponent
Tin standard solution, 1000 mg/L Sn in dil. nitric acid with hydrofluoric acid (max. 1%) standard for ICP
Tin standard solution, 1000 mg/L Sn in dil. nitric acid with hydrofluoric acid (max. 1%) standard for ICP
Catalog # ACCUICP63N5
Supplier:  AccuStandard
CAS Number:  
Tin standard solution, 1000 mg/L Sn in dil. nitric acid with hydrofluoric acid (max. 1%) standard for ICP
Catalog # ACCUICP63N5
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Tin
  • Symbol:
    Sn

Specifications

About this item