Order Entry
Belgium
ContactUsLinkComponent
Silicon standard solution, 1,000 mg/L Si in water with hydrofluoric acid (max. 1%) standard for ICP-MS
Silicon standard solution, 1,000 mg/L Si in water with hydrofluoric acid (max. 1%) standard for ICP-MS
Catalog # ACCUICPMS52W01X1
Supplier:  AccuStandard
CAS Number:  
Silicon standard solution, 1,000 mg/L Si in water with hydrofluoric acid (max. 1%) standard for ICP-MS
Catalog # ACCUICPMS52W01X1
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O/tr HF
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Silicon
  • Symbol:
    Si

Specifications

About this item