Phosphorus standard solution, 10,000 mg/L P in water standard for ICP
Catalog # REGEPP4B7
Supplier: REAGECON
CAS Number:
Specifications
- Concentration:10000mg/L
- Matrix:H₂O
- Pack type:Glass bottle
- Pk:250 mL
- Environmentally Preferable:
- Element:Phosphorus
- Symbol:P