Order Entry
Belgium
ContactUsLinkComponent
Phosphorus standard solution, 1,000 mg/L P in water standard for AAS
Phosphorus standard solution, 1,000 mg/L P in water standard for AAS
Catalog # REGEAAP-H
Supplier:  REAGECON
CAS Number:  
Phosphorus standard solution, 1,000 mg/L P in water standard for AAS
Catalog # REGEAAP-H
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Phosphorus
  • Symbol:
    P

Specifications

About this item