Order Entry
Belgium
ContactUsLinkComponent
Copper standard solution, 1000 mg/L Cu in dil. nitric acid standard for ICP
Copper standard solution, 1000 mg/L Cu in dil. nitric acid standard for ICP
Catalog # REGEPCU2A2
Supplier:  REAGECON
CAS Number:  
Copper standard solution, 1000 mg/L Cu in dil. nitric acid standard for ICP
Catalog # REGEPCU2A2
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2-5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Copper
  • Symbol:
    Cu

Specifications

About this item