Copper standard solution, 1,000 mg/L Cu in 0,5 M nitric acid standard for AAS
Catalog # REGEAACUH
Supplier: REAGECON
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:0,5 M HNO₃
- Pack type:Glass bottle
- Pk:500 mL
- Element:Copper
- Symbol:Cu