Order Entry
Belgium
ContactUsLinkComponent
Potassium standard solution, 1,000 mg/L K in dilute nitric acid standard for AAS
Potassium standard solution, 1,000 mg/L K in dilute nitric acid standard for AAS
Catalog # ACRO196205000
CAS Number:  
Potassium standard solution, 1,000 mg/L K in dilute nitric acid standard for AAS
Catalog # ACRO196205000
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2-5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Potassium
  • Symbol:
    K

Specifications

About this item