Order Entry
Belgium
ContactUsLinkComponent
 
Iron standard solution, 10000 mg/L Fe in dil. nitric acid standard for ICP-MS
  ACCUICPMS27N1
 :  
Iron standard solution, 10000 mg/L Fe in dil. nitric acid standard for ICP-MS
  ACCUICPMS27N1
 :  

 

  • Concentration:
    10000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Element:
    Iron
  • Symbol:
    Fe