Order Entry
Belgium
ContactUsLinkComponent
 
Iron standard solution, 1000 mg/L Fe in dil. nitric acid standard for ICP
Catalog # REGEPFE2A2
Supplier:  REAGECON
CAS Number:  
Iron standard solution, 1000 mg/L Fe in dil. nitric acid standard for ICP
Catalog # REGEPFE2A2
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Iron
  • Symbol:
    Fe

Specifications

About this item