Order Entry
Belgium
ContactUsLinkComponent
J.T.Baker®, Calcium Standard Solution, 1000 mg/L Ca in dil. Nitric Acid (from Ca) Standard for AAS
J.T.Baker®, Calcium Standard Solution, 1000 mg/L Ca in dil. Nitric Acid (from Ca) Standard for AAS
Catalog # 6925.0100
Supplier:  Avantor
CAS Number:  
J.T.Baker®, Calcium Standard Solution, 1000 mg/L Ca in dil. Nitric Acid (from Ca) Standard for AAS
Catalog # 6925.0100
Supplier:  Avantor
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Calcium
  • Symbol:
    Ca

Specifications

About this item

Certifications: Traceable to NIST