ContactUsLinkComponent
Potassium chloride saturated with silver chloride aqueous solution 4 mol/l (4 N) in aqueous solution for filling electrodes
  KRAF20116.4000
 :  
Potassium chloride saturated with silver chloride aqueous solution 4 mol/l (4 N) in aqueous solution for filling electrodes
  KRAF20116.4000
 :  
  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096
 :
 :74.55 g/mol
 :1.13 g/cm³ (20 °C)
 :Ambient
 :231-211-8
 :7447-40-7
 :KCl+AgCl
 :MFCD00011360