Order Entry
Belgium
ContactUsLinkComponent
Zirconium standard solution, 1000 mg/L Zr in dil. nitric acid standard for ICP
Zirconium standard solution, 1000 mg/L Zr in dil. nitric acid standard for ICP
Catalog # ACCUICP71N5
Supplier:  AccuStandard
CAS Number:  
Zirconium standard solution, 1000 mg/L Zr in dil. nitric acid standard for ICP
Catalog # ACCUICP71N5
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Zirconium
  • Symbol:
    Zr

Specifications

About this item