Order Entry
Belgium
ContactUsLinkComponent
Tungsten standard solution, 1000 mg/L W in water (from (NH₄)₂WO₄) Certipur® standard for ICP, Supelco®
Tungsten standard solution, 1000 mg/L W in water (from (NH₄)₂WO₄) Certipur® standard for ICP, Supelco®
Catalog # 1.70364.0100
Supplier:  Merck
CAS Number:  
undefined
Tungsten standard solution, 1000 mg/L W in water (from (NH₄)₂WO₄) Certipur® standard for ICP, Supelco®
Catalog # 1.70364.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Tungsten
  • Symbol:
    W

Specifications

About this item

Certifications: Traceable to SRM from NIST