Order Entry
Belgium
ContactUsLinkComponent
Specpure® Tungsten Standard Solution, 1000 mg/L W in Water (from (NH₄)₂WO₄) Standard for ICP
Specpure® Tungsten Standard Solution, 1000 mg/L W in Water (from (NH₄)₂WO₄) Standard for ICP
Certificates
Specpure® Tungsten Standard Solution, 1000 mg/L W in Water (from (NH₄)₂WO₄) Standard for ICP
Certificates

Specifications

  • MDL No.:
    MFCD00011461
  • Physical form:
    Liquid
  • Molecular formula:
    (NH4)2WO4 in H2O

Specifications

Related Information

Frequently Bought Together