ContactUsLinkComponent
Rhodium standard solution, 1000 mg/L Rh in 20% hydrochloric acid (from RhCl₃), standard for ICP, Specpure®
Rhodium standard solution, 1000 mg/L Rh in 20% hydrochloric acid (from RhCl₃), standard for ICP, Specpure®
Certificates
Rhodium standard solution, 1000 mg/L Rh in 20% hydrochloric acid (from RhCl₃), standard for ICP, Specpure®
Certificates
Specifications
  • MDL No.:
    MFCD00011201