Order Entry
Belgium
ContactUsLinkComponent
Dimethyl sulphoxide ≥99.9%, ChromAR® for HPLC, Macron Fine Chemicals™
Dimethyl sulphoxide ≥99.9%, ChromAR® for HPLC, Macron Fine Chemicals™
Catalog # 2969-25
   Safer Human and Environmental Health
CAS Number:  
undefined
Dimethyl sulphoxide ≥99.9%, ChromAR® for HPLC, Macron Fine Chemicals™
Catalog # 2969-25
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

Specifications

Specification Test Results

  • ACS - Assay ((CH₃)₂SO) (by GC, corrected for water):
    ≥99.9 %
  • ACS - Residue after Evaporation:
    ≤5 ppm
  • ACS - Titrable Acid (meq/g):
    ≤0.001
  • ACS - Water (H₂O)(by KF, coulometric):
    ≤0.05 %
  • Optical Absorbance (1-cm path vs water) - 270 nm:
    ≤0.40
  • Optical Absorbance (1-cm path vs water) - 290 nm:
    ≤0.18
  • Optical Absorbance (1-cm path vs water) - 310 nm:
    ≤0.06
  • Optical Absorbance (1-cm path vs water) - 330 nm:
    ≤0.02
  • Optical Absorbance (1-cm path vs water) - 350 - 400 nm:
    ≤0.01

Specification Test Results

About this item

 :
 :78.14 g/mol
Merck Index:13,03285
Boiling Pt:189 °C (1013 hPa)
Melting Pt:18.45 °C
Density:1.101 g/cm³ (20 °C)
Flash Pt:89 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-664-3
CAS Number:67-68-5
Formula:(CH₃)₂SO
MDL Number:MFCD00002089