Sodium standard solution, 1000 mg/L Na in dil. nitric acid standard for AAS
Supplier: AccuStandard
Certificates
About this item
Frequently Bought Together
Methanol, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Acetonitrile, anhydrous (max. 0.001 % H₂O)
Supplier: Merck
Methanol, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Ethyl acetate 99.5% for analysis
Supplier: Thermo Fisher Scientific
Toluene (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
Supplier: PanReac AppliChem
N-Methyl-2-pyrrolidone (NMP) ≥99.7%, Biotechnology Grade, Burdick & Jackson™
Supplier: Honeywell Chemicals
N,N-Dimethylacetamide, anhydrous (max. 0.005% H₂O)
Supplier: VWR Chemicals
n-Hexane, extra dry 96+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
2-Propanol solution in water 70%, technical
Supplier: Thermo Fisher Scientific