Order Entry
Belgium
ContactUsLinkComponent
Sodium standard solution, 1,000 mg/L Na in 0.5 M nitric acid standard for AAS
Sodium standard solution, 1,000 mg/L Na in 0.5 M nitric acid standard for AAS
Catalog # ACRO196311000
CAS Number:  
Sodium standard solution, 1,000 mg/L Na in 0.5 M nitric acid standard for AAS
Catalog # ACRO196311000
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Sodium
  • Symbol:
    Na

Specifications

About this item