Order Entry
Belgium
ContactUsLinkComponent
 
Molybdenum standard solution, 1000 mg/L Mo in water standard for AAS
Catalog # REGEAAMOH
Supplier:  REAGECON
CAS Number:  
Molybdenum standard solution, 1000 mg/L Mo in water standard for AAS
Catalog # REGEAAMOH
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Molybdenum
  • Symbol:
    Mo

Specifications

About this item