Order Entry
Belgium
ContactUsLinkComponent
Lithium standard solution, 1000 mg/L Li in 0,5 M nitric acid standard for AAS
Lithium standard solution, 1000 mg/L Li in 0,5 M nitric acid standard for AAS
Catalog # REGEAALIH
Supplier:  REAGECON
CAS Number:  
Lithium standard solution, 1000 mg/L Li in 0,5 M nitric acid standard for AAS
Catalog # REGEAALIH
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Lithium
  • Symbol:
    Li

Specifications

About this item