Gold standard solution, 1,000 mg/L Au in 10% hydrochloric acid standard for ICP-MS
Catalog # ACCUICPMS22H01X1
Supplier: AccuStandard
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:10% HCl
- Pack type:Glass bottle
- Pk:100 mL
- Element:Gold
- Symbol:Au