Germanium standard solution, 1,000 mg/L Ge in water with hydrofluoric acid (max. 1%) standard for AAS
Catalog # ACCUAA21W1
Supplier: AccuStandard
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:H₂O/tr HF
- Pack type:Glass bottle
- Pk:100 mL
- Environmentally Preferable:
- Element:Germanium
- Symbol:Ge