Silicone Viscosoty Standards for Rotational Viscometers
CANNON silicone viscosity standards are intended for use with rotational viscometers only.
Silicone viscosity standards are not recommended for glass capillary viscometers (including vacuum viscometers), metal cup-type viscometers (such as ISO, Zahn, Ford and Shell cups), or any other viscometers providing kinematic viscosity—CANNON hydrocardon-based oil standards should be used for these types of viscometers.
Viscosity standards are traceable to a NIST calibration and prepared in Cannon's ISO 9001-registered, A2LA-accredited laboratory.
: Supplied with certificate.
: Viscosities quoted in the table below are nominal. Lot specific viscosity data in mPa·s (cP) at 20 °C, 23 °C, 24 °C, 25 °C, 26 °C, 27 °C and 40 °C is provided with each standard.
Frequently Bought Together