Magnesium standard solution, 1000 mg/L Mg in dil. nitric acid standard for ICP-MS
Catalog # ACCUICPMS32N0.1X1
Supplier: AccuStandard
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Glass bottle
- Pk:100 mL
- Element:Magnesium
- Symbol:Mg