Order Entry
Belgium
ContactUsLinkComponent
Chromium standard solution, 1,000 mg/L Cr in dil. nitric acid standard for ICP
Chromium standard solution, 1,000 mg/L Cr in dil. nitric acid standard for ICP
Catalog # REGEPCR2A2
Supplier:  REAGECON
CAS Number:  
Chromium standard solution, 1,000 mg/L Cr in dil. nitric acid standard for ICP
Catalog # REGEPCR2A2
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2-5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Chromium
  • Symbol:
    Cr

Specifications

About this item