Order Entry
Belgium
ContactUsLinkComponent
Manganese standard solution, 1000 mg/L Mn in dil. nitric acid standard for ICP
Manganese standard solution, 1000 mg/L Mn in dil. nitric acid standard for ICP
Catalog # REGEPMN2A2
Supplier:  REAGECON
CAS Number:  
Manganese standard solution, 1000 mg/L Mn in dil. nitric acid standard for ICP
Catalog # REGEPMN2A2
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Manganese
  • Symbol:
    Mn

Specifications

About this item