Order Entry
Belgium
ContactUsLinkComponent
 
Storage vials, without caps, Chromacol™
  548-0687
 :  CHROMACOL
undefined
Storage vials, without caps, Chromacol™
  548-0687
 :  CHROMACOL

 

  • Cap included:
    No
  • Ø×H:
    15×46 mm
  • Capacity:
    3,50 ml
  • Colour:
    Clear
  • Description:
    Vials with high recovery base profile
  • Material:
    Galvanised iron/ceramic
  • Pk:
    250

 

 

These vials are manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) glass and feature a 13-425 thread finish.