Sampling scoops, WHEATON®
240 mm, Acier inoxydable
Catalog # WHEA885540
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
This scoop is made from 302 stainless steel and is ideal for sampling powders, slurries, soils, and more.
Specifications
- Longueur:240 mm
- Matière:Acier inoxydable