Gas Washing Bottle, Head with VitraPOR® Filter Disc, Standard Joint NS 45/40
About this item
ROBU® gas washing bottle head with integrated VitraPOR® filter disc and standard ground joint ensures efficient gas-liquid contact and uniform dispersion in absorption processes.
High-quality borosilicate glass construction
- Includes a VitraPOR® filter disc for efficient filtration of gases
- Standard joint for easy connection to other laboratory equipment NS 45/40
- Resistant to chemicals and high temperatures
- Ideal for gas washing and purification
- Reusable and easy to clean
- Ensures reliable filtration and separation of impurities
- Compatible with standard laboratory setups
The ROBU gas washing bottle head with VitraPOR filter disc and standard joint is designed for efficient gas scrubbing in laboratory setups. The integrated filter disc ensures fine dispersion of gases into the washing liquid, maximising surface contact and absorption efficiency. Made from high-quality borosilicate glass, it offers excellent chemical resistance and durability. The standardised joint ensures a secure, leak-free fit with compatible gas washing bottles. Ideal for removing impurities, neutralising aggressive gases, or humidifying gas streams, the ROBU head component guarantees reliable performance in chemical processing and analytical applications.
Certifications: In compliance with ISO 4796-80 and ASTM E128-99 standards.
3 Options Available Below
Product Details & Documents
ROBU® gas washing bottle head with integrated VitraPOR® filter disc and standard ground joint ensures efficient gas-liquid contact and uniform dispersion in absorption processes.
High-quality borosilicate glass construction
- Includes a VitraPOR® filter disc for efficient filtration of gases
- Standard joint for easy connection to other laboratory equipment NS 45/40
- Resistant to chemicals and high temperatures
- Ideal for gas washing and purification
- Reusable and easy to clean
- Ensures reliable filtration and separation of impurities
- Compatible with standard laboratory setups
The ROBU gas washing bottle head with VitraPOR filter disc and standard joint is designed for efficient gas scrubbing in laboratory setups. The integrated filter disc ensures fine dispersion of gases into the washing liquid, maximising surface contact and absorption efficiency. Made from high-quality borosilicate glass, it offers excellent chemical resistance and durability. The standardised joint ensures a secure, leak-free fit with compatible gas washing bottles. Ideal for removing impurities, neutralising aggressive gases, or humidifying gas streams, the ROBU head component guarantees reliable performance in chemical processing and analytical applications.
Certifications: In compliance with ISO 4796-80 and ASTM E128-99 standards.
Documents
Product Family Options
Product Information
Head for washing bottle
Product Information
- Catalog #PorosityDescriptionFor bottlesNSExt.ØAvailabilityPrice
Accessories
Product Information
- Catalog #CapacityNSAvailabilityPrice
- Washing bottle lower part