Order Entry
Belgium
ContactUsLinkComponent
 
Centrifuge, manually operated
  523-4491
 :  Hettich
   Low Manufacturing Impact
undefined
Centrifuge, manually operated
  523-4491
 :  Hettich
   Low Manufacturing Impact

 

  • Description:
    Manually-operated centrifuge
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)
  • Pk:
    1

 

 

This convenient centrifuge can be easily clamped to a bench or table and operated without electricity.

  • Self-lubricating gear (maintenance-free)
  • Can be operated with two different rotors: 1014 and 1025
  • Rotor 1014 can accelerate 15 ml conical tubes up to 1300 ×g
  • Rotor 1025 accomodates various conical and round 15 ml tubes (maximum 17×125 mm)
 : Rotors and tubes available separately.

Rotors