Verso™ qRT-PCR Kits
Supplier: Thermo Fisher Scientific
About this item
The Verso™ qRT-PCR system combines the Verso™ RT enzyme with ABsolute™ Blue qPCR master mix to provide maximum convenience and versatility. Available in a 1- or 2-step format, as well as multiple pack sizes, to suit all application and throughput requirements.
- The combining of Verso™ enzyme and Thermo-Start™ Taq polymerase gives sensitive results from low copy number RNA template
- Inert blue dye for easy visualisation of the qRT-PCR reaction
- No need to perform an RNase H digestion after cDNA synthesis, therefore streamlining the reaction protocol
- RT enhancer saves time by eliminating the need for DNase treatment of samples
- Multiple priming options for optimisation of specific RNA templates
12 Options Available Below
Product Details & Documents
The Verso™ qRT-PCR system combines the Verso™ RT enzyme with ABsolute™ Blue qPCR master mix to provide maximum convenience and versatility. Available in a 1- or 2-step format, as well as multiple pack sizes, to suit all application and throughput requirements.
- The combining of Verso™ enzyme and Thermo-Start™ Taq polymerase gives sensitive results from low copy number RNA template
- Inert blue dye for easy visualisation of the qRT-PCR reaction
- No need to perform an RNase H digestion after cDNA synthesis, therefore streamlining the reaction protocol
- RT enhancer saves time by eliminating the need for DNase treatment of samples
- Multiple priming options for optimisation of specific RNA templates
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 733-1112
- 733-1114
- 733-1117
- 733-1119
- 733-1122
- 733-1124
- 733-1127
- 733-1129
- 733-1132
- 733-1134
- 733-1137
- 733-1139
- Application:
- RT-qPCR dye
- RT-qPCR dye
- RT-qPCR dye
- RT-qPCR dye
- RT-qPCR dye
- RT-qPCR dye
- RT-qPCR dye
- RT-qPCR dye
- RT-qPCR dye
- RT-qPCR dye
- RT-qPCR dye
- RT-qPCR dye
- One step:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
Specifications
Product Family Options
Product Information
- PkAvailabilityPrice
- 733-1112Verso™ 1-Step qRT-PCR Kit plus ROX vial, 200×25 µl reactionsVerso™ 1-Step qRT-PCR Kit plus ROX vial, 200×25 µl reactions1 KITLog in for availability1 * 1 KIT926,00 €Specifications:
More
Specifications
Cat. No.733-1112ApplicationRT-qPCR dyeOne stepYes - 733-1114Verso™ 1-Step qRT-PCR Kit plus ROX vial, 400×25 µl reactionsVerso™ 1-Step qRT-PCR Kit plus ROX vial, 400×25 µl reactions1 KITLog in for availability1 * 1 KIT1 750,00 €Specifications:
More
Specifications
Cat. No.733-1114ApplicationRT-qPCR dyeOne stepYes - 733-1117Verso™ 1-Step qRT-PCR ROX Kit, 200×25 µl reactionsSpecifications:
More
Specifications
Cat. No.733-1117ApplicationRT-qPCR dyeOne stepYes - 733-1119Verso™ 1-Step qRT-PCR ROX Kit, 400×25 µl reactionsSpecifications:
More
Specifications
Cat. No.733-1119ApplicationRT-qPCR dyeOne stepYes - 733-1122Verso™ 1-Step qRT-PCR Low ROX Kit, 200×25 µl reactionsVerso™ 1-Step qRT-PCR Low ROX Kit, 200×25 µl reactions1 KITLog in for availability1 * 1 KIT932,00 €Specifications:
More
Specifications
Cat. No.733-1122ApplicationRT-qPCR dyeOne stepYes - 733-1124Verso™ 1-Step qRT-PCR Low ROX Kit, 400×25 µl reactionsVerso™ 1-Step qRT-PCR Low ROX Kit, 400×25 µl reactions1 KITLog in for availability1 * 1 KIT1 750,00 €Specifications:
More
Specifications
Cat. No.733-1124ApplicationRT-qPCR dyeOne stepYes - 733-1127Verso™ SYBR® Green 1-Step qRT-PCR Kit plus ROX vial, 200×25 µl reactionsVerso™ SYBR® Green 1-Step qRT-PCR Kit plus ROX vial, 200×25 µl reactions1 KITLog in for availability1 * 1 KIT1 050,00 €Specifications:
More
Specifications
Cat. No.733-1127ApplicationRT-qPCR dyeOne stepYes - 733-1129Verso™ SYBR® Green 1-Step qRT-PCR Kit plus ROX vial, 400×25 µl reactionsVerso™ SYBR® Green 1-Step qRT-PCR Kit plus ROX vial, 400×25 µl reactions1 KITLog in for availability1 * 1 KIT2 070,00 €Specifications:
More
Specifications
Cat. No.733-1129ApplicationRT-qPCR dyeOne stepYes - 733-1132Verso™ SYBR® Green 1-Step qRT-PCR ROX Kit, 200×25 µl reactionsVerso™ SYBR® Green 1-Step qRT-PCR ROX Kit, 200×25 µl reactions1 KITLog in for availability1 * 1 KIT1 040,00 €Specifications:
More
Specifications
Cat. No.733-1132ApplicationRT-qPCR dyeOne stepYes - 733-1134Verso™ SYBR® Green 1-Step qRT-PCR ROX Kit, 400×25 µl reactionsVerso™ SYBR® Green 1-Step qRT-PCR ROX Kit, 400×25 µl reactions1 KITLog in for availability1 * 1 KIT2 080,00 €Specifications:
More
Specifications
Cat. No.733-1134ApplicationRT-qPCR dyeOne stepYes - 733-1137Verso™ SYBR® Green 1-Step qRT-PCR Low ROX Kit, 200×25 µl reactionsVerso™ SYBR® Green 1-Step qRT-PCR Low ROX Kit, 200×25 µl reactions1 KITLog in for availability1 * 1 KIT1 070,00 €Specifications:
More
Specifications
Cat. No.733-1137ApplicationRT-qPCR dyeOne stepYes - 733-1139Verso™ SYBR® Green 1-Step qRT-PCR Low ROX Kit, 400×25 µl reactionsVerso™ SYBR® Green 1-Step qRT-PCR Low ROX Kit, 400×25 µl reactions1 KITLog in for availability1 * 1 KIT2 110,00 €Specifications:
More
Specifications
Cat. No.733-1139ApplicationRT-qPCR dyeOne stepYes