RT-PCR kits, Verso™
The Verso™ RT-PCR system combines a reverse transcription enzyme, improved priming options and optimised buffers to generate high yield and full length cDNA. With a variety of kit options for both 1- and 2-step RT-PCR reactions, Verso™ provides optimum flexibility and sensitivity in an easy to use format.
- Application:RT-qPCR dye
- One step:Yes
Frequently Bought Together