Order Entry
Belgium
ContactUsLinkComponent
VWR® Phosphate Buffered Saline (PBS), High Phosphate
VWR® Phosphate Buffered Saline (PBS), High Phosphate
Catalog # 444037E
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® Phosphate Buffered Saline (PBS), High Phosphate
Catalog # 444037E
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Bag-in-box
  • Pk:
    10 L
  • Description:
    Phosphate Buffered Saline (PBS), High Phosphate

Specifications

About this item

Irradiated solution, pH 7,2, for washing blots.

  • NaCl 8,77 g/L
  • Na₂HPO₄, 12H₂0 19,2 g/L
  • NaH₂PO₄, 2H₂O 6,55 g/L