Standards for 2100N and 2100N IS
Supplier: Hach
Certificates
Frequently Bought Together
Quinine sulphate dihydrate, TECHNICAL
Supplier: VWR Chemicals
Syringe filters, Acrodisc®, Nylon
Supplier: Cytiva (Formerly Pall Lab)
α-Methylstyrene ≥99.0% (by GC) stabilised
Supplier: TCI
Orthoboric acid 4% ± 0.05% in aqueous solution, Fluka™
Supplier: Honeywell Chemicals
Bottle-top dispensers, Dispensette® S Organic
Supplier: Brand
Saccharin, Supelco®
Supplier: Merck
Butyl acetate, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
D-(-)-Aspartic acid ≥99%
Supplier: Thermo Fisher Scientific
Caps for 20 ml glass crimp top headspace vials, level 1 everyday analysis, SureSTART™
Supplier: Thermo Fisher Scientific
VitraPOR® Glass Filter Discs, Standard
Supplier: ROBU GLASFILTER
Conductivity sensors
Supplier: Mettler - Toledo
Replacement parts for GC packed injectors
Supplier: PerkinElmer
m-Cymene ≥99.0% (by GC)
Supplier: TCI
Turbidity calibration standards, formazin, Supelco®
Supplier: Merck
Turbidity standards, Stablcal®
Supplier: Hach
Molecular sieve 3A (0.3 nm, 3 Å) 4 --> 8 mesh
Supplier: Thermo Fisher Scientific
α-Methylstyrene ≥99% stabilised
Supplier: Thermo Fisher Scientific