Buffer solutions in sachets
About this item
Solutions in sachets ensure optimum precision.
- Advantages
- No danger of contamination
The plastic-coated sachet provides a perfect liquid seal and good protection from light. The solutions should only be used once to avoid any danger of contamination. They also satisfy the requirements of the GLP and ISO standards. They have been adjusted using a pH-metre that has been calibrated with a buffer associated with NIST standards.
Delivery information: The buffers in sachets are delivered with a certificate of conformity.
6 Options Available Below
Product Details & Documents
Solutions in sachets ensure optimum precision.
- Advantages
- No danger of contamination
The plastic-coated sachet provides a perfect liquid seal and good protection from light. The solutions should only be used once to avoid any danger of contamination. They also satisfy the requirements of the GLP and ISO standards. They have been adjusted using a pH-metre that has been calibrated with a buffer associated with NIST standards.
Delivery information: The buffers in sachets are delivered with a certificate of conformity.
Documents
Product Family Options
Product Information
- Reference temperaturePack typePk InfoPkAvailabilityPrice
- 662-0540Buffer solutions 4,01/7,00/9,21
- 662-0544Buffer solutions 4,01/7,00/10,01 (blend)Buffer solutions 4,01/7,00/10,01 (blend)25 °CSachet30×20 ml30Log in for availability1 * 30 ST68,70 €
- 662-0538Buffer solution 7,00
- 662-0541Buffer solution 4,01
- 662-0542Buffer solution 9,21
- 662-0543Buffer solution 10,01