Order Entry
Belgium
ContactUsLinkComponent
Corning® T-Cell Culture Supplement with ConA (IL-2 Culture Supplement), Corning
Corning® T-Cell Culture Supplement with ConA (IL-2 Culture Supplement), Corning
Catalog # 734-1310
Supplier:  Corning
   Low Manufacturing Impact
CAS Number:  
Corning® T-Cell Culture Supplement with ConA (IL-2 Culture Supplement), Corning
Catalog # 734-1310
Supplier:  Corning
   Low Manufacturing Impact
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Source:
    Splenocyte culture
  • Species:
    Rat
  • Description:
    T-Cell Culture Supplement with ConA, rat
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 - Bedford
  • Formulation:
    Conditioned medium (RPMI 1640) from ConA-stimulated rat splenocyte cultures containing 10% foetal bovine serum and 10 mM HEPES.  ConA content is 10 µg/ml of medium.
  • Quality control:
    Tested for ability to promote growth of an IL-2 dependent mouse cell line (CTLL) in serum-containing medium
    Filtered (0,2 µm membrane) and tested and found negative for bacteria, fungi and mycoplasma
  • Storage and stability:
    Stable for at least three months at 2 to 8 °C

Specifications

About this item

T-STIM with ConA (IL-2 culture supplement), rat, is used to promote proliferation and activation of T-cells and support high-titer HIV production by leukocytes.

Media used to promote proliferation and activation (PHA or ConA Supplemented Media) of T-cells and support high-titer HIV production by leukocytes.

Tested for ability to promote proliferation of an IL-2 dependent mouse cell line (CTLL) in serum-containing medium. Filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi, and mycoplasma.